Insta-Check pH Papers
pH paper jumbo dispenser Insta-Check, 15 ft., ⁵⁄₃₂", 0 - 13
Catalog # 470203-358
Supplier: Micro Essential
Supplier #: 3015
Jump to:
Product Details & Documents
The Ultimate in Wide Range, General Purpose pH Papers.
- One dip determines any pH from 0 - 13
- Distinct and easily read color separations
Insta-Check 0 - 13 provides 14 brilliant colors on 1 paper giving instant pH checks over the entire range with accurate, reproducible readings; and immediate response. Available in different widths to meet your special requirements and preferences. Paper rolls come with dispenser; refills do not.
Specifications
- Width:⁵⁄₃₂"
- Description:pH paper jumbo dispenser Insta-Check
- Length:15 ft.
- pH range:0 - 13