Orders LinkContactUsLinkComponent
Phosphate, Low Range
Phosphate, Low Range
LaMotte® Water Test Kit, Phosphate, Low Range
Catalog # 470203-078
Supplier:   LaMotte
Supplier #:  3121-02
Phosphate, Low Range
Catalog # 470203-078
Supplier:  LaMotte
Phosphate, Low Range
Phosphate, Low Range
LaMotte® Water Test Kit, Phosphate, Low Range
Price
193,20 $
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab. 
  • Range: 0–2 ppm
  • Material for 50 Tests
  • Compact, Waterproof Carrying Case Contains Kit
  • Instructions Included
  • Pre-calibrated Test Tubes
Specifications
  • Description:
    LaMotte® Water Test Kit, Phosphate, Low Range
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.