Order Entry
Canada
Orders LinkContactUsLinkComponent
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

1000 ml, 8
Catalog # 470202-160
Supplier:   Corning
Supplier #:  4060-1L
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Catalog # 470202-160
Supplier:  Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

1000 ml, 8

Price
$52.70
Log in for availability
Each Case of 6
Product Details & Documents

Manufactured from borosilicate glass.

Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).

Documents

Specifications
  • Capacity:
    1000 ml
  • Stopper no.:
    8
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.