Orders LinkContactUsLinkComponent
Parafilm
Parafilm
Parafilm, Roll Size, in. x ft.: 2 x 250
  470201-930
Supplier #:  HS234526A
Parafilm
  470201-930
Parafilm
Parafilm
Parafilm, Roll Size, in. x ft.: 2 x 250
$64.60
Each Case of 24
Flexible, self-adhesive, waterproof plastic for closures, mounting and more.
Adheres quickly and firmly to itself, covering culture tube or flask mouths and sealing petri dish edges. Excellent resistance to air, other gases, alcohol, common acids, and moderately concentrated alkalies.
Documents
  • Description:
    Parafilm, Roll Size, in. x ft.: 2 x 250