Disposable Polypropylene Snap Cap Test Tubes
Disposable Polypropylene Snap Cap Test Tubes, 17 x 100 mm, 14 mL Cap
Supplier: Corning
Supplier #: 352059
Jump to:
Product Details & Documents
Ensured sterility with the convenience of disposability makes these tubes ideal for all types of laboratory procedures.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Disposable Polypropylene Snap Cap Test Tubes, 17 x 100 mm, 14 mL Cap
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



