Orders LinkContactUsLinkComponent
Document Analysis Lab Activity
  470200-744
Document Analysis Lab Activity
  470200-744
 :  
Document Analysis Lab Activity
Document Analysis Lab Activity
Determine a document’s authenticity.
  • Grades 6-12
  • Analysis of secret messages using chemistry
Your students will be fascinated as they perform both physical and chemical analysis of handwriting materials found at a crime scene. The procedures they will follow cover checking for erasures, abnormal positioning of a signature, use of different inks, and differences in handwriting or typing. It comes with enough materials for six setups and an instruction manual. A UV lamp, required, is available separately.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.