Order Entry
Canada
Orders LinkContactUsLinkComponent
Wool Fire Blanket with Rectangular Wall Case

Wool Fire Blanket with Rectangular Wall Case

84" L x 66" W
Catalog # 470200-728
Wool Fire Blanket with Rectangular Wall Case
Catalog # 470200-728
Supplier:  
Wool Fire Blanket with Rectangular Wall Case
Wool Fire Blanket with Rectangular Wall Case

84" L x 66" W

Discontinued
Product Details & Documents

These blankets are 100% flame resistant.

  • For smothering fires
  • Also serves as first aid blanket
  • Wool fabrication
  • Wall case includes bracket for easy accessibility
  • Blankets have 100% flame resistance

This wool blanket can be used to smother fires or as a first aid blanket.

The wall case can be easily attached to lab walls for accessibility during emergencies.

Specifications
  • Dimensions:
    84" L x 66" W