Fingerprint Analysis Lab Activity
Le produit 470002-508 est un remplacement de 470200-096 et a le même emballage et les mêmes spécifications, mais peut être d'une quantité différente.
Product Details & Documents
A fun and educational introduction to criminalistics.
- Grades 7-14
- Multiple methods for detecting latent prints
Your students will answer the question “Whodunit?” while learning how to dust and develop latent fingerprints. They “lift” sets of prints from various surfaces, turning them over for others to analyze. After comparing the prints to standards developed by the FBI, and working with “Modus Operandi” worksheets, the class will identify the suspects. The kit includes instructor’s and assessment guides, student data sheets, and all materials but the fingerprints for 24 students.
Spécifications
- Description:Fingerprint Analysis Kit
- No. of lab groups:6
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)