Certipur® pH Buffer Solutions, MilliporeSigma
Boric acid/potassium chloride/sodium hydroxide solution, Borate, 25 °C, Sachet, 30×30 ml, 9.00, Colorless
Supplier: MilliporeSigma
: 1.99003.0001
Certipur® pH Buffer Solutions, MilliporeSigma
Catalog # CA1.99003.0001
Supplier: MilliporeSigma
CAS Number:
Jump to:
Product Details & Documents
Traceable to SRM from NIST and PTB.
Certificates
Search by Lot Number1.07827.1000_08072012 1.09400.0500_08162012 1.09400.4000_08162012 1.09400.9010_08162012 1.09407.0500_08022012
More
Specifications
Specifications
- Pack type:Sachet
- Color:Colorless
- Description:Boric acid/potassium chloride/sodium hydroxide solution
- Size:30×30 ml
- pH buffer type:Borate
- Reference temperature:25 °C
- pH:9.00
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



