Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Certipur® pH Buffer Solutions, MilliporeSigma

Di-sodium hydrogen phosphate/potassium di-hydrogen phosphate, Phosphate, 20 °C, Plastic bottle, 1 L, 7.00, Colorless
  CA1.09439.1000
 :  1.09439.1000
Certipur® pH Buffer Solutions, MilliporeSigma
  CA1.09439.1000
 :  
Certipur® pH Buffer Solutions, MilliporeSigma
Certipur® pH Buffer Solutions, MilliporeSigma

Di-sodium hydrogen phosphate/potassium di-hydrogen phosphate, Phosphate, 20 °C, Plastic bottle, 1 L, 7.00, Colorless

 :  
 
$147.71
   
Each (1l) Case of 6 (1l)
 
   
 

 

  • Pack type:
    Plastic bottle
  • Color:
    Colorless
  • Description:
    Di-sodium hydrogen phosphate/potassium di-hydrogen phosphate
  • Size:
    1 L
  • pH buffer type:
    Phosphate
  • Reference temperature:
    20 °C
  • pH:
    7.00

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.