Order Entry
Canada
Orders LinkContactUsLinkComponent
Standard-Grade Glass Beaker

Standard-Grade Glass Beaker

1000 ml, 100 ml, 50 - 1000 ml
Catalog # 470191-202
Supplier:   VWR International
Standard-Grade Glass Beaker
Catalog # 470191-202
Supplier:  VWR International
Standard-Grade Glass Beaker
Standard-Grade Glass Beaker

1000 ml, 100 ml, 50 - 1000 ml

Price
$27.20
Log in for availability
Each Case of 24
Product Details & Documents

Our high-quality borosilicate beakers meet ASTM standards and are a great value. With uniform wall thickness, white enamel, double-graduated metric scale, and a marking spot. Beakers can be heated and used with acids, salt solutions, and organic solvents.

Documents

Specifications
  • Graduation Interval:
    100 ml
  • Graduation Range:
    50 - 1000 ml
  • Capacity:
    1000 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.