Orders LinkContactUsLinkComponent
Standard-Grade Glass Beaker
10 ml (0.34 oz.)
  470191-186
Standard-Grade Glass Beaker
  470191-186
Standard-Grade Glass Beaker
Standard-Grade Glass Beaker
10 ml (0.34 oz.)
$8.85
Each Case of 48
Our high-quality borosilicate beakers meet ASTM standards and are a great value. With uniform wall thickness, white enamel, double-graduated metric scale, and a marking spot. Beakers can be heated and used with acids, salt solutions, and organic solvents.
  • Capacity:
    10 ml (0.34 oz.)