Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Plastic Beads

Plastic Beads, 8 mm
Catalog # 470023-956
Supplier:   Avantor
Plastic Beads
Catalog # 470023-956
Supplier:  Avantor
Plastic Beads
Plastic Beads

Plastic Beads, 8 mm

Price
16,45 $
Log in for availability
Pack of 100
Product Details & Documents

Packages of approximately 100.

  • 4 mm Red bead = Acrylic
  • 8 mm Navy bead = Acrylic
  • 12 mm White bead = Acetate
  • Autoclavable
Specifications
  • Ø:
    8 mm
  • Description:
    Plastic Beads
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.