pH Test Papers
pH Test Papers, Litmus Red ACS (12 Vials)
Catalog # 470123-118
Supplier: Precision Laboratories
Supplier #: 190-12V-100
Jump to:
Product Details & Documents
Each stoppered vial contains 100 (1.8 x 2") strips.
Each stoppered vial contains 100 (1.8" x 2") strips.
Specifications
- Description:pH Test Papers, Litmus Red ACS (12 Vials)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



