Boreal Standard Compound Microscopes
Product Details & Documents
Boreal standard microscopes feature metal construction, a rugged student-proof design, and precision optics making them a favorite among educators.
- Coated-glass optics
- Wide-field eyepiece with pointer
- Limited lifetime warranty
These microscopes optics are parfocal and parcentered for a crisp, consistent field of view. All units feature strategically located controls, course and fine focus, and a 360° rotatable head. Units are available in a variety of configurations offering three or four objectives, plain or mechanical stage, and corded or cordless charging.
Ordering information: Microscopes include a manual, dust cover, and limited warranty.
Specifications
- Description:Compound microscope
- Stage:Mechanical
- Condenser type:Iris
- Illumination:LED cordless
- Cat. No.:470014-588
- Objectives:4×, 10×, 40×
Accessories
Product Information
- Catalog #ElectricalAvailabilityPrice
- Replacement bulb
Product Information
- Catalog #ElectricalAvailabilityPrice
- Replacement Bulb, with Bayonet Base