Orders LinkContactUsLinkComponent
Curved Microdissection Forceps
Curved Microdissection Forceps
Straight Microdissection Forceps, Stainless steel, 115 mm (4.5"), Straight
Catalog # 470007-556
Supplier:   Dunrite Instruments
Supplier #:  6853305
Curved Microdissection Forceps
Catalog # 470007-556
Curved Microdissection Forceps
Straight Microdissection Forceps, Stainless steel, 115 mm (4.5"), Straight
Price
$16.20
Log in for availability
Each
Product Details & Documents
Micro fine, curved or straight points enhance control when probing the most delicate structures.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Description:
    Straight Microdissection Forceps
  • Length:
    115 mm (4.5")
  • Material:
    Stainless steel
  • Tip style:
    Straight