Orders LinkContactUsLinkComponent
Hook Collars
Hook Collars
Hook Collar, Fits 19 mm (0.75") Rod
Catalog # 470006-702
Supplier #:  HKCL02
Hook Collars
Catalog # 470006-702
Hook Collars
Hook Collars
Hook Collar, Fits 19 mm (0.75") Rod
Price
$21.95
Log in for availability
Each
Product Details & Documents
Quickly add a hook to a support rod.
  • Plated steel
  • Thumbscrew
Plated steel collar clamps feature thumbscrews and hooks for suspending pulleys, balances, etc. from support rods or crossbars.
Specifications
  • Description:
    Hook Collar, Fits 19 mm (0.75") Rod