Orders LinkContactUsLinkComponent
Balls
Balls
Hardwood ball, drilled, ¾"
Catalog # 470137-644
Supplier:   Wards
Supplier #:  HWBL19
Balls
Catalog # 470137-644
Supplier:  Wards
Balls
Balls
Hardwood ball, drilled, ¾"
Clearance
$3.45
Sale
Valid until 31/12/2026
Log in for availability
Each
Product Details & Documents
Useful for collision and pendulum experiments.
A must have in any physics classroom or laboratory.
Specifications
  • Description:
    Hardwood ball, drilled
  • Diameter:
    ¾"
  • Cat. No.:
    470137-644