Orders LinkContactUsLinkComponent
Ward's® Limestone (Shell) | Avantor
Ward's® Limestone (Shell)
Limestone (Shell), hand specimen, 2×2 - 3×4"
Catalog # 470026-084
Supplier:   Avantor
Ward's® Limestone (Shell)
Catalog # 470026-084
Supplier:  Avantor
Ward's® Limestone (Shell)
Ward's® Limestone (Shell)
Limestone (Shell), hand specimen, 2×2 - 3×4"
Price
$32.05
Log in for availability
Each
Product Details & Documents
Buff, coarse, crystalline “Austin Marble” with shell casts and molds.
  • Collected in Texas
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Specifications
  • Description:
    Limestone (Shell), hand specimen
  • Specimen dimensions:
    2×2 - 3×4"