Orders LinkContactUsLinkComponent
Tectonics Model
Tectonics Model
Tectonics model, complete lab, —, —, —
Catalog # 470004-622
Supplier:   Avantor
Tectonics Model
Catalog # 470004-622
Supplier:  Avantor
Tectonics Model
Tectonics Model
Tectonics model, complete lab, —, —, —
Price
$269.05
Log in for availability
Each Pallet of 36
Product Details & Documents
This Tectonics Model demonstrates the principle of superposition and the concepts of transgression, regression, and compression.
  • Provides dramatic visualization of faults
  • Students calculate as they recreate tectonic compression
  • Instructions included
  • Teacher developed
  • Aligned to NGSS standards (MS/HS: ESS1, ESS2)
Add a new quantitative dimension to tectonics demonstrations with this innovative device. When using this box, pressure is applied with a screw mechanism that measures equal increments of force over distance. Students can simulate folding, faulting, and mountain building.
Accessories information: Replacement sediments available.
Specifications
  • Description:
    Tectonics model, complete lab
  • Height:
  • Length:
  • Width:
Recommendations