Orders LinkContactUsLinkComponent
Ward's® Cell Manipulative Models
Ward's® Cell Manipulative Models
Cell Manipulative Models, Plant, 10
Catalog # 470003-988
Supplier:   Avantor
Ward's® Cell Manipulative Models
Catalog # 470003-988
Supplier:  Avantor
Ward's® Cell Manipulative Models
Ward's® Cell Manipulative Models
Cell Manipulative Models, Plant, 10
Price
$139.80
Log in for availability
Kit of 1
Product Details & Documents
Students Create Their Own Cells
Introduce, reinforce, and test your students’ knowledge of cell structures and their functions with these fun and easy to use manipulative models. Using Velcro, students can attach organelles to the cell as they discover the fascinating complexity of plant and animal cells. Each model set comes with ten laminated cell sheets and ten sets of organelles, as well as a teacher’s guide with suggested activities.
Specifications
  • Description:
    Cell Manipulative Models, Plant
  • No. of lab groups:
    10