Wire Gauzes, Plain
Wire Gauze, Plain, 5" x 5", 13×13 cm (5×5")
Catalog # 470188-996
Supplier: United Scientific Supplies
: WGPL55
Jump to:
Product Details & Documents
Sturdy wire mesh for supporting glassware over a Bunsen burner.
Long lasting, heat- and tear-resistant iron
- Available in a variety of sizes
Sheet of cross-hatched wire used to support glassware during heating. Galvanized iron wire provides a long-life expectancy and is resistant to tearing over time. Edges are folded over for a clean perimeter surface.
Specifications
Specifications
- Description:Wire Gauze, Plain, 5" x 5"
- Size:13×13 cm (5×5")
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



