Order Entry
Canada
Orders LinkContactUsLinkComponent
Ward's® Plasmid DNA for Transformations

Ward's® Plasmid DNA for Transformations

pUC19 WITHOUT Loading Dye, 1 µg/100 µl
Catalog # 470184-902
Supplier:   Avantor
Ward's® Plasmid DNA for Transformations
Catalog # 470184-902
Supplier:  Avantor
CAS Number:  
Ward's® Plasmid DNA for Transformations
Ward's® Plasmid DNA for Transformations

pUC19 WITHOUT Loading Dye, 1 µg/100 µl

CAS Number:  
Price
$29.25
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

These plasmids allow you to perform bacterial transformations, conferring new antibiotic resistance, as a visual way of demonstrating gene transfer.

  • Purchase only the amount of plasmid you need
  • Many plasmids already contain loading dye for added convenience

Plasmid pUC19: 2686 base pairs. Confers antibiotic resistance to ampicillin and complements defects in b-galactosidase in appropriate bacterial strains. Isolated from E. coli JM101.

pBR322: 4361 base pairs. Commonly used plasmid cloning vector for E. coli, and confers resistance to Ampicillin and Tetracycline.

Ordering information: These products are perishable materials, and are shipped next-day air on ice to ensure freshness. Please specify a delivery date at order time.

Specifications

Specifications

  • Concentration:
    1 µg/100 µl
  • Description:
    pUC19 WITHOUT Loading Dye

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.