Tooth Development Slides
Dentine formation slide, Section, Mallory Triple
Catalog # 470182-806
Supplier: Avantor
Jump to:
Product Details & Documents
Fetal pig tissue showing early tooth development.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Slide samples created at various stages of mammal tooth-development.
Specifications
- Description:Dentine formation slide
- Stain:Mallory Triple
- Orientation:Section