Selaginella, Strobilus Slide
Catalog # 470181-148
Supplier: Avantor
Specifications
- Description:Selaginella, Strobilus
- Cat. no.:470181-148
- Stain:Quadruple
- Supplier No.:WARD470181-148
- Orientation:Longitudinal section
Specifications
About this item
Spikemoss
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows micro- and megasporophylls.
Frequently Bought Together