Orders LinkContactUsLinkComponent
Lambda Markers
Lambda Markers
Lambda DNA/EcoR I Hind III Digest
Catalog # 470175-932
Supplier:   Avantor
Lambda Markers
Catalog # 470175-932
Supplier:  Avantor
CAS Number:  
Lambda Markers
Lambda Markers
Lambda DNA/EcoR I Hind III Digest
CAS Number:  
Price
77,35 $
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
These DNA markers are used to determine size of unknown DNA fragments. If using within 2–3 days, they do not need to be refrigerated and can be stored right on a shelf. If storing for longer time periods, freezing is recommended.
Ship date is required.
Specifications
Specifications
  • Description:
    Lambda DNA/EcoR I Hind III Digest