Orders LinkContactUsLinkComponent
Lambda Markers
Lambda Markers
Lambda Phage DNA
Catalog # 470175-926
Supplier:   Avantor
Lambda Markers
Catalog # 470175-926
Supplier:  Avantor
CAS Number:  
Lambda Markers
Lambda Markers
Lambda Phage DNA
CAS Number:  
Price
$64.70
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
These DNA markers are used to determine size of unknown DNA fragments. If using within 2–3 days, they do not need to be refrigerated and can be stored right on a shelf. If storing for longer time periods, freezing is recommended.
Ship date is required.
Specifications
Specifications
  • Description:
    Lambda Phage DNA
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.