Orders LinkContactUsLinkComponent
Ward's® Forensics Slide Sets
General Forensics Slide Set
  470177-066
Ward's® Forensics Slide Sets
  470177-066
 :  
Ward's® Forensics Slide Sets
Ward's® Forensics Slide Sets
General Forensics Slide Set
Students view the identifying features of a crime scene in detail.
  • Easy-to-Distinguish Identifying Features
  • Rigorous Quality Control Standards
  • Three Different Sets Available
The slide sets include all the evidence needed to solve the crime.
The sets meet rigorous quality control standards.
 :
The general forensics set includes blood, human (sm); blood, mammal (sm); bloodstained textile fibers (wm); cheek cells, human (sm); DNA in cells (sect); fingerprints, three types (wm); hair, three types (wm); skin, human (sect).
The trace evidence set includes air pollution (wm); cellulose paper fiber (wm); chalk (wm); corn starch (wm); cosmetic types (sm); dandruff (wm); dust, volcanic (wm); dust, common (wm); dust, iron (wm); fiberglass (wm); pollen, mixed, sixteen types (wm); pollen, mixed, twenty types (wm); pollen, mature, lily (wm);pollen, germinating (wm); salt (wm); sand (wm); sawdust (wm); tobacco, dried (wm); toothpaste (sm).
The fiber types set includes cotton/linen/hemp/silk(wm); nylon/Dacron/rayon (wm); dyed fibers(wm); nylon/ wool(wm); silk(wm); wool(wm).
  • Description:
    General Forensics Slide Set