Balls
Whiffle ball, 2.5", Package of 6
Catalog # 470173-904
Supplier: SPORT SUPPLY GROUP, INC.
Supplier #: 426306
Jump to:
Product Details & Documents
Useful for collision and pendulum experiments.
A must have in any physics classroom or laboratory.
Specifications
- Description:Whiffle ball
- Packaging:Package of 6
- Diameter:2.5"