Order Entry
Canada
Orders LinkContactUsLinkComponent
Plastic Slide Box for 25 Slides

Plastic Slide Box for 25 Slides

Plastic Slide Box, 25, 5.5 x 3.5 x 1.3"
Catalog # 470156-718
Supplier:   C & A Scientific
Supplier #:  97-0025
Plastic Slide Box for 25 Slides
Catalog # 470156-718
Supplier:  C & A Scientific
Plastic Slide Box for 25 Slides
Plastic Slide Box for 25 Slides

Plastic Slide Box, 25, 5.5 x 3.5 x 1.3"

Price
$12.15
Log in for availability
Each Case of 200
Product Details & Documents

Store your slides efficiently.

  • Store your slides safely and keep them well organized

The plastic box protects 1" x 3" slides and won’t warp when the temperature and humidity vary. It is stackable, with telescoping covers, a cork bottom, and embossed slide numbers on the bottom.

Specifications
  • Capacity:
    25
  • Description:
    Plastic Slide Box
  • Dimensions:
    5.5 x 3.5 x 1.3"