Polyethylene Disposable Gloves
Polyethylene gloves, Medium, Box/100
Catalog # 470153-722
Supplier: Dival Safety & Supplies
Supplier #: GDPEMED
Jump to:
Product Details & Documents
Ultra sensitive.
- Ambidextrous
- Powder-Free
These disposable, polyethylene gloves feature sheer-gauge 1.25 mil, which permits near bare hand sense of touch while protecting hands and labware from contamination. Gloves are powder-free, so they will not mark delicate materials.
Specifications
- Glove size:Medium
- Description:Polyethylene gloves
- Packaging:Box/100
- Powdered/Powder-free:Powder-free
- Cat. No.:470153-722
- Disposable / reusable:Disposable
- Ambidextrous/Hand-specific:Ambidextrous