Precision pH Test Strips
pH paper strips, 11.5–14.0
Catalog # 470151-004
Supplier: Precision Laboratories
Supplier #: PH1114-1B-50
Jump to:
Product Details & Documents
The indicator on these strips won’t contaminate the solutions the strips are testing.
- Vinyl-backed
- Non-contaminating indicator
- Fifty strips per package
Durable pH testing strips that are backed with vinyl.
Specifications
- Description:pH paper strips
- pH range:11.5–14.0