Passer à En-tête
Passer à Contenu principal
Passer à Pied de page
Avancé
Avancé
SDS
Certificats
Données du fabricant
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
UNIVERSAL INDICATOR 2X30ML
470109-614
:
Ward's Science
UNIVERSAL INDICATOR 2X30ML
470109-614
:
Ward's Science
:
UU0010-PK2
ghs_470109-614.pdf
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International