Sharp-Point Surgical Scissors
Sharp-Point Surgical Scissors, 115 mm (4.5"), Stainless steel, Sharp/Sharp
Catalog # 470106-340
Supplier: Dunrite Instruments
Supplier #: 140980
Jump to:
Product Details & Documents
Start your cuts easily.
- Dissect without a scalpel
- Handles rounded for ambidextrous use
- Available in two sizes
Sharp-pointed, straight surgical scissors allow the user to insert blades into tissue to start the cut. Larger scissors provide greater leverage for tough cutting; shorter scissors for finer work. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Description:Sharp-Point Surgical Scissors
- Length:115 mm (4.5")
- Material:Stainless steel
- Blade type:Sharp/Sharp
Accessories
Product Information
- Catalog #LengthMaterialAvailabilityPrice
- Surgical Scissors
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



