Slide Storage Boxes
Slide Box, Wood with Plastic Inserts, 100, 24 x 19 x 3 cm
Catalog # 470104-374
Supplier: United Scientific Supplies
Supplier #: WSB100
Jump to:
Product Details & Documents
Excellent For Storing Standard Sized Slides
- Keep slides debris-free
- Store slides neatly and efficiently
Each slide slips into a separate, numbered compartment for easy reference.
Specifications
- Capacity:100
- Description:Slide Box, Wood with Plastic Inserts
- Dimensions:24 x 19 x 3 cm