Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Triangular Files

Triangular file, File: 9", 11"
  470104-232
 :  TRF006
Triangular Files
  470104-232
Triangular Files
Triangular Files

Triangular file, File: 9", 11"

 
17,20 $
   
Each
 

Useful for metal filing or glass cutting.

  • Wooden handle for easy gripping
  • Hardened triple sides

Triangular file has a tapered shape, and is mounted on a hardwood handle for easy gripping. For general laboratory use, including to score glass before cutting.

 
  • Description:
    Triangular file
  • Length:
    File: 9"
  • Overall length:
    11"
Accessories

 

  •  
    Description
    Length
    Overall Length
     
     
  • 470005-474
    Triangular file
    File: 4"
    7"
       
    Each - $12.65