Miscellaneous Convenience Items
Spring, Compressible 2 1/2 x 3/4''
Catalog # 470092-392
Supplier: STANLEY SPRING & STAMPING
: BAGGED
Jump to:
Product Details & Documents
: WARNING: CHOKING HAZARD. Not for children under 3 years.
Specifications
- Description:Spring, Compressible 2 1/2 x 3/4''
Accessories
Product Information
- Catalog #AvailabilityPrice
- Funnel, Poly, 114 mL (4 oz)
Product Information
- Catalog #AvailabilityPrice
- Single sheave
Product Information
- Catalog #AvailabilityPrice
- Ball-Bearing Clamp Pulley
Product Information
- Catalog #AvailabilityPrice
- Battery Holder, 9 V
Product Information
- Catalog #AvailabilityPrice
- Weight, Hooked, 10 g to 1000 g, Cast Iron, Set/9Weight, Hooked, 10 g to 1000 g, Cast Iron, Set/9470003-584Log in for availabilityKit of 1 - $130.05
Product Information
- Catalog #ColorLengthAvailabilityPrice
- Braided Nylon pulley cord, tensile strength 200 lbs. testBraided Nylon pulley cord, tensile strength 200 lbs. test470005-414White9 m (10 yds.)Log in for availabilityEach - $31.00
Product Information
- Catalog #AvailabilityPrice
- Assorted Plastic Pulleys



