Solid Balls for Physics Experiments
Solid Steel Ball, 10 mm, ³/₈", -
Catalog # 470089-322
Supplier: BUFFALO BEARINGS INC.
Supplier #: 2754800
Jump to:
Product Details & Documents
Drilled balls are used for pendulum bobs or impact experiments, while our other balls are suitable for collision apparatus and density/volume experiments.
Caution: WARNING: CHOKING HAZARD -Item contains a small ball. Not for children under 3 years.
Specifications
- Description:Solid Steel Ball, 10 mm
- Packaging:-
- Diameter:³/₈"