Orders LinkContactUsLinkComponent
Precision pH Test Strips
Precision pH Test Strips
pH paper strips, 0.0–1.5
Catalog # 470150-922
Precision pH Test Strips
Catalog # 470150-922
Supplier:  
Precision pH Test Strips
Precision pH Test Strips
pH paper strips, 0.0–1.5
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
STRIPS PH INDICATOR PH0-2.5 SPCL 100STI Catalog #EM1.09540.0001
Product EM1.09540.0001 is similar to 470150-922 but manufacturer and specifications may vary.
Product Details & Documents
The indicator on these strips won’t contaminate the solutions the strips are testing.
  • Vinyl-backed
  • Non-contaminating indicator
  • Fifty strips per package
Durable pH testing strips that are backed with vinyl.
Specifications
  • Description:
    pH paper strips
  • pH range:
    0.0–1.5