Right Angle Prisms
Right Angle Prisms, Acrylic, 36 x 25 mm
Catalog # 470149-476
Supplier: Shiv Dial Sud & Sons
Supplier #: PH30307
Jump to:
Product Details & Documents
For Versatile Optical Applications
- These prisms are polished on all faces
- They are made of shatter-resistant acrylic material or optical quality glass
These prisms are polished on all faces. They are made of shatter-resistant acrylic material or optical quality glass. The length of the hypotenuse and the prism height are given for each item.
Specifications
- Description:Right Angle Prisms, Acrylic, 36 x 25 mm