VWR® Florence Boiling Flasks
125 ml, 2
Supplier: VWR International
Jump to:
Product Details & Documents
High Quality Borosilicate Glass
- Meet ASTM standards
- Round body and long neck are ideal for boiling liquids without excessively rapid evaporation
: Rubber stoppers are not included.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:125 ml
- Rubber stopper size:2