Orders LinkContactUsLinkComponent
VWR® Florence Boiling Flasks
VWR® Florence Boiling Flasks
1000 ml, 8
Catalog # 470149-284
Supplier:   VWR International
VWR® Florence Boiling Flasks
Catalog # 470149-284
Supplier:  VWR International
VWR® Florence Boiling Flasks
VWR® Florence Boiling Flasks
1000 ml, 8
Price
43,60 $
Log in for availability
Each Case of 6
Product Details & Documents
High Quality Borosilicate Glass
  • Meet ASTM standards
  • Round body and long neck are ideal for boiling liquids without excessively rapid evaporation
Ordering information: Rubber stoppers are not included.
Documents
Specifications
  • Capacity:
    1000 ml
  • Rubber stopper size:
    8