Order Entry
Canada
Orders LinkContactUsLinkComponent
 

VWR® Test Tubes

5 ml, 000, 12×75 mm
Catalog # 470149-272
Supplier:   VWR International
VWR® Test Tubes
Catalog # 470149-272
Supplier:  VWR International
VWR® Test Tubes
VWR® Test Tubes

5 ml, 000, 12×75 mm

Price
$100.35
Log in for availability
Pack of 72 Case of 720
Product Details & Documents

VWR® Test tubes are made from high-quality glass.

  • Borosilicate glass construction
  • Uniform thickness

Each tube features a strong lip.

 : ASTM standards.

 

Specifications
  • Capacity:
    5 ml
  • Stopper size:
    000
  • O.D.×L:
    12×75 mm