Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Test Tubes

VWR® Test Tubes

9 ml, 000, 13×100 mm
Catalog # 470149-270
Supplier:   VWR International
VWR® Test Tubes
Catalog # 470149-270
Supplier:  VWR International
VWR® Test Tubes
VWR® Test Tubes

9 ml, 000, 13×100 mm

Price
$102.55
Log in for availability
Pack of 72 Case of 720
Product Details & Documents

VWR® Test tubes are made from high-quality glass.

  • Borosilicate glass construction
  • Uniform thickness

Each tube features a strong lip.

Certifications: ASTM standards.

Documents

Specifications
  • Capacity:
    9 ml
  • Stopper size:
    000
  • O.D.×L:
    13×100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.