Orders LinkContactUsLinkComponent
Lil'pet Micropipette
Lil'pet Micropipette
100 µl
Catalog # 470148-872
Supplier:   VWR International
Lil'pet Micropipette
Catalog # 470148-872
Supplier:  VWR International
Lil'pet Micropipette
Lil'pet Micropipette
100 µl
Price
$70.00
Log in for availability
Each Case of 100
Product Details & Documents
Lil'pet Micropipettes feature ideal volumes for gel loading.
  • Units accept standard micropipette tips
  • Economical
  • Durable
  • Autoclavable
These fixed-volume pipettes feature a double stop plunger to expel all liquid from the tip.
The 1000 μl model includes two tips. All other pipettes come with one each.
Specifications
  • Volume:
    100 µl