Order Entry
Canada
Orders LinkContactUsLinkComponent
Polypropylene Jars

Polypropylene Jars

Polypropylene Jars, 500 ml
Catalog # 470147-474
Supplier #:  421155
Polypropylene Jars
Catalog # 470147-474
Supplier:  
Polypropylene Jars
Polypropylene Jars

Polypropylene Jars, 500 ml

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
JAR POLYPROPYLENE WHITE 16OZ 89MM Catalog #470178-178

Product 470178-178 is a functional alternative to 470147-474 but Packaging, Quantity, Manufacturer and/or specifications may vary.

Product Details & Documents

Convenient for Storing and Collecting Lab Supplies and Specimens.

  • Supplied with linerless polypropylene screw caps
  • Leak-proof
  • Autoclavable
Specifications
  • Capacity:
    500 ml
  • Description:
    Polypropylene Jars
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.