Orders LinkContactUsLinkComponent
Polypropylene Jars
Polypropylene Jars
Polypropylene Jars, 250 ml
Catalog # 470147-472
Supplier:   Dynalon
Supplier #:  421145
Polypropylene Jars
Catalog # 470147-472
Supplier:  Dynalon
Polypropylene Jars
Polypropylene Jars
Polypropylene Jars, 250 ml
Price
$174.40
Log in for availability
Pack of 6 Case of 36
Product Details & Documents
Convenient for Storing and Collecting Lab Supplies and Specimens.
  • Supplied with linerless polypropylene screw caps
  • Leak-proof
  • Autoclavable
Specifications
  • Capacity:
    250 ml
  • Description:
    Polypropylene Jars