Polypropylene Beakers
400 ml
Catalog # 470145-342
Supplier: Bel-Art
Supplier #: F26214-0000
Jump to:
Product Details & Documents
Translucent Polypropylene Beakers.
- Raised graduations ±5% accuracy
- Stack without sticking together
- Autoclavable up to 121°C
Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents.
Specifications
- Capacity:400 ml



