Order Entry
Canada
Orders LinkContactUsLinkComponent
Polypropylene Beakers

Polypropylene Beakers

4000 ml
Catalog # 470145-340
Supplier:   Bel-Art
Supplier #:  F26219-0000
Polypropylene Beakers
Catalog # 470145-340
Supplier:  Bel-Art
Polypropylene Beakers
Polypropylene Beakers

4000 ml

Price
$93.60
Log in for availability
Each Case of 6
Product Details & Documents

Translucent Polypropylene Beakers.

  • Raised graduations ±5% accuracy
  • Stack without sticking together
  • Autoclavable up to 121°C

Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents.

Specifications
  • Capacity:
    4000 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.