Orders LinkContactUsLinkComponent
Glass Stirring Rods
3 mm, 127 mm
  470144-862
 :  GSR005-PK/144
Glass Stirring Rods
  470144-862
Glass Stirring Rods
Glass Stirring Rods
3 mm, 127 mm
$12.00
Pack of 12 Case of 144
Glass stirring rod features fire polished flint glass.
  • Rounded ends
Stirring rods are made of soft flint glass, with both ends fire-polished and rounded.
  • Ø:
    3 mm
  • Length:
    127 mm